Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
-
Chromosome Location
Chr.19: 45995457 - 46018616 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267723.1 NP_001254652.1 10 - 11 1289 100
XM_005259191.3 XP_005259248.1 10 - 11 3394 100
XM_005259192.4 XP_005259249.1 10 - 11 2035 100
XM_011527257.2 XP_011525559.1 10 - 11 1318 100
GenBank mRNA CA775840.1 - 2 - 3 227 100
CB852675.1 - 3 - 4 276 100
GQ375050.1 - 6 - 7 722 100

View Full Product Details