Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
-
Chromosome Location
Chr.19: 45995457 - 46018616 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267723.1 NP_001254652.1 3 - 4 282 90
XM_005259191.3 XP_005259248.1 3 - 4 2387 90
XM_005259192.4 XP_005259249.1 3 - 4 1028 90
XM_011527257.2 XP_011525559.1 3 - 4 311 90
GenBank mRNA DR007040.1 - 3 - 4 302 90
GQ375050.1 - 3 - 4 255 90

View Full Product Details