Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC61
Entrez Gene ID
729440
Gene Name
coiled-coil domain containing 61
Gene Aliases
-
Chromosome Location
Chr.19: 45995457 - 46018616 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267723.1 NP_001254652.1 5 - 6 601 73
XM_005259191.3 XP_005259248.1 5 - 6 2706 73
XM_005259192.4 XP_005259249.1 5 - 6 1347 73
XM_011527257.2 XP_011525559.1 5 - 6 630 73
GenBank mRNA AW453013.1 - 3 - 4 73
BI819372.1 - 4 - 5 501 73

View Full Product Details