The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267723.1 | NP_001254652.1 | 9 - 10 | 1148 | 134 |
XM_005259191.3 | XP_005259248.1 | 9 - 10 | 3253 | 134 | |
XM_005259192.4 | XP_005259249.1 | 9 - 10 | 1894 | 134 | |
XM_011527257.2 | XP_011525559.1 | 9 - 10 | 1177 | 134 | |
GenBank mRNA | CA775840.1 | - | 1 - 2 | 86 | 134 |
CB852675.1 | - | 2 - 3 | 135 | 134 |