The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198465.3 | NP_940867.2 | 6 - 7 | 786 | 116 |
XM_006724632.3 | XP_006724695.1 | 6 - 7 | 748 | 116 | |
XM_006724633.3 | XP_006724696.1 | 6 - 7 | 748 | 116 | |
XM_006724634.3 | XP_006724697.1 | 6 - 7 | 748 | 116 | |
XM_011530887.2 | XP_011529189.1 | 6 - 7 | 540 | 116 | |
GenBank mRNA | AK131549.1 | - | 6 - 7 | 748 | 116 |
BX538345.1 | - | 6 - 7 | 788 | 116 |