The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175854.7 | NP_787050.6 | 3 - 4 | 771 | 66 |
XM_005266332.3 | XP_005266389.1 | 3 - 4 | 821 | 66 | |
XM_005266333.3 | XP_005266390.1 | 3 - 4 | 619 | 66 | |
XM_005266334.1 | XP_005266391.1 | 2 - 3 | 181 | 66 | |
XM_011535032.2 | XP_011533334.1 | 3 - 4 | 765 | 66 | |
XM_011535033.2 | XP_011533335.1 | 3 - 4 | 765 | 66 | |
XM_011535034.2 | XP_011533336.1 | 3 - 4 | 539 | 66 | |
XM_017020528.1 | XP_016876017.1 | 3 - 4 | 765 | 66 | |
XM_017020529.1 | XP_016876018.1 | 3 - 4 | 254 | 66 | |
XM_017020530.1 | XP_016876019.1 | 3 - 4 | 824 | 66 | |
XM_017020532.1 | XP_016876021.1 | 3 - 4 | 313 | 66 | |
XM_017020533.1 | XP_016876022.1 | 3 - 4 | 712 | 66 | |
GenBank mRNA | AB107584.1 | - | 2 - 3 | 181 | 66 |
AB109552.1 | - | 2 - 3 | 181 | 66 | |
AK296435.1 | - | 3 - 4 | 771 | 66 | |
BC128179.1 | - | 2 - 3 | 181 | 66 | |
BC128180.1 | - | 2 - 3 | 181 | 66 |