The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010855.3 | NP_001010855.1 | 6 - 7 | 632 | 67 |
NR_110865.1 | 6 - 7 | 884 | 67 | ||
XM_011523677.2 | XP_011521979.1 | 7 - 8 | 645 | 67 | |
XM_011523678.2 | XP_011521980.1 | 7 - 8 | 644 | 67 | |
XM_011523680.2 | XP_011521982.1 | 7 - 8 | 645 | 67 | |
XM_011523681.2 | XP_011521983.1 | 6 - 7 | 893 | 67 | |
XM_011523683.2 | XP_011521985.1 | 4 - 5 | 330 | 67 | |
XM_017024228.1 | XP_016879717.1 | 7 - 8 | 644 | 67 | |
XM_017024229.1 | XP_016879718.1 | 7 - 8 | 644 | 67 | |
XM_017024230.1 | XP_016879719.1 | 7 - 8 | 644 | 67 | |
XM_017024231.1 | XP_016879720.1 | 3 - 4 | 821 | 67 | |
GenBank mRNA | AK091819.1 | - | 6 - 7 | 886 | 67 |
AY753192.1 | - | 4 - 5 | 392 | 67 | |
BC125169.1 | - | 1 - 2 | 36 | 67 |