The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001010855.3 | NP_001010855.1 | 10 - 11 | 1128 | 68 |
NM_001290211.1 | NP_001277140.1 | 10 - 11 | 1496 | 68 | |
NR_110865.1 | 9 - 10 | 1223 | 68 | ||
XM_011523677.2 | XP_011521979.1 | 11 - 12 | 1141 | 68 | |
XM_011523680.2 | XP_011521982.1 | 11 - 12 | 1141 | 68 | |
XM_011523681.2 | XP_011521983.1 | 10 - 11 | 1389 | 68 | |
XM_011523683.2 | XP_011521985.1 | 8 - 9 | 826 | 68 | |
XM_017024228.1 | XP_016879717.1 | 11 - 12 | 1140 | 68 | |
XM_017024229.1 | XP_016879718.1 | 11 - 12 | 1140 | 68 | |
XM_017024230.1 | XP_016879719.1 | 11 - 12 | 1140 | 68 | |
XM_017024231.1 | XP_016879720.1 | 7 - 8 | 1317 | 68 | |
GenBank mRNA | AK091819.1 | - | 9 - 10 | 1225 | 68 |
AY753192.1 | - | 8 - 9 | 888 | 68 | |
BC110301.1 | - | 10 - 11 | 1479 | 68 | |
BC125169.1 | - | 5 - 6 | 532 | 68 |