Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMPD3
Entrez Gene ID
84443
Gene Name
FERM and PDZ domain containing 3
Gene Aliases
-
Chromosome Location
Chr.X: 107522450 - 107605264 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032428.1 NP_115804.1 8 - 9 744 92
XM_006724709.2 XP_006724772.1 2 - 3 219 92
XM_011531056.2 XP_011529358.1 6 - 7 720 92
XM_011531057.2 XP_011529359.1 6 - 7 646 92
XM_011531058.2 XP_011529360.1 5 - 6 558 92
XM_017029901.1 XP_016885390.1 6 - 7 965 92
XM_017029902.1 XP_016885391.1 6 - 7 965 92

View Full Product Details