Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYRM5
Entrez Gene ID
144363
Gene Name
LYR motif containing 5
Gene Aliases
-
Chromosome Location
Chr.12: 25195216 - 25205051 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005253320.4 XP_005253377.1 1 - 2 125 107

View Full Product Details