Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD50
Entrez Gene ID
57182
Gene Name
ankyrin repeat domain 50
Gene Aliases
-
Chromosome Location
Chr.4: 124664048 - 124712732 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020337.2 NP_065070.1 1 - 2 273 101
GenBank mRNA AK291068.1 - 1 - 2 273 101
CX871714.1 - 1 - 2 154 101

View Full Product Details