The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128215.1 | NP_001121687.1 | 4 - 5 | 744 | 81 |
XM_011539748.2 | XP_011538050.1 | 4 - 5 | 715 | 81 | |
XM_011539751.2 | XP_011538053.1 | 3 - 4 | 4084 | 81 | |
XM_011539752.2 | XP_011538054.1 | 2 - 3 | 84 | 81 | |
XM_011539753.2 | XP_011538055.1 | 4 - 5 | 715 | 81 | |
GenBank mRNA | BC157888.1 | - | 5 - 6 | 712 | 81 |
BC171908.1 | - | 5 - 6 | 712 | 81 | |
EF426484.1 | - | 4 - 5 | 734 | 81 |