The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171183.1 | NP_001164654.1 | 2 - 3 | 287 | 66 |
XM_017009350.1 | XP_016864839.1 | 2 - 3 | 772 | 66 | |
GenBank mRNA | AI024830.1 | - | 2 - 3 | 66 | |
CN445899.1 | - | 2 - 3 | 296 | 66 |