Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NKX1-2
Entrez Gene ID
390010
Gene Name
NK1 homeobox 2
Gene Aliases
C10orf121, NKX-1.1, bB238F13.2
Chromosome Location
Chr.10: 124447429 - 124449981 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146340.1 NP_001139812.1 1 - 2 250 143

View Full Product Details