Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMD1
Entrez Gene ID
90378
Gene Name
sterile alpha motif domain containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 14088417 - 14090420 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC007384.2 - 3 - 4 441 78
BC030129.2 - 3 - 4 620 78
BC065477.1 - 3 - 4 588 78
BC080588.1 - 3 - 4 441 78

View Full Product Details