Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SPINK13
- Entrez Gene ID
-
153218
- Gene Name
- serine peptidase inhibitor, Kazal type 13 (putative)
- Gene Aliases
-
HBVDNAPTP1, HESPINTOR, LiESP6, SPINK5L3
- Chromosome Location
-
Chr.5: 148268860 - 148286210 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK001520.1 |
- |
2 - 3 |
498 |
82 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AU129688.1 |
- |
2 - 3 |
498 |
|
Chr. 5 - 148268180 - 148282204 [+] on Build GRCh38 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105378220 |
LOC105378220 |
|
-24.7kb
|
Sense |
Chr. - 148238298 - 148245526 on Build GRCh38 |
Downstream |
serine peptidase inhibitor, Kazal type 7 (putative) |
SPINK7 |
ECG2, ECRG2 |
42.3kb
|
Sense |
Chr. - 148312427 - 148315919 on Build GRCh38 |
View Full Product Details