The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001040129.2 | NP_001035218.1 | 4 - 5 | 299 | 117 |
GenBank mRNA | AK001520.1 | - | 5 - 6 | 768 | 117 |
BC128163.1 | - | 5 - 6 | 451 | 117 | |
BC128164.1 | - | 4 - 5 | 299 | 117 | |
EF108324.1 | - | 4 - 5 | 347 | 117 | |
EF426754.1 | - | 4 - 5 | 347 | 117 |