The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272046.1 | NP_001258975.1 | 4 - 5 | 589 | 65 |
NM_001320804.1 | NP_001307733.1 | 4 - 5 | 531 | 65 | |
XM_011539757.2 | XP_011538059.1 | 4 - 5 | 861 | 65 | |
XM_017016177.1 | XP_016871666.1 | 4 - 5 | 341 | 65 | |
XM_017016179.1 | XP_016871668.1 | 4 - 5 | 294 | 65 | |
XM_017016181.1 | XP_016871670.1 | 4 - 5 | 338 | 65 | |
GenBank mRNA | AJ536328.2 | - | 4 - 5 | 513 | 65 |
AK122716.1 | - | 4 - 5 | 584 | 65 | |
AK127756.1 | - | 4 - 5 | 420 | 65 | |
AY572972.1 | - | 4 - 5 | 351 | 65 | |
BC143380.1 | - | 4 - 5 | 291 | 65 |