The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291661.1 | NP_001278590.1 | 14 - 15 | 1953 | 137 |
XM_011533466.1 | XP_011531768.1 | 14 - 15 | 1950 | 137 | |
XM_011533467.1 | XP_011531769.1 | 14 - 15 | 1917 | 137 | |
XM_011533468.1 | XP_011531770.1 | 14 - 15 | 1950 | 137 | |
XM_011533469.1 | XP_011531771.1 | 14 - 15 | 1950 | 137 | |
XM_011533471.2 | XP_011531773.1 | 14 - 15 | 1950 | 137 | |
XM_011533474.1 | XP_011531776.1 | 14 - 15 | 1950 | 137 | |
XM_017005860.1 | XP_016861349.1 | 14 - 15 | 1950 | 137 | |
XM_017005861.1 | XP_016861350.1 | 13 - 14 | 2633 | 137 |