The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080423.3 | NP_001073892.3 | 20 - 21 | 2611 | 87 |
XM_011534141.2 | XP_011532443.1 | 20 - 21 | 2693 | 87 | |
XM_011534142.2 | XP_011532444.1 | 20 - 21 | 2605 | 87 | |
XM_011534143.2 | XP_011532445.1 | 19 - 20 | 2567 | 87 | |
XM_011534145.2 | XP_011532447.1 | 14 - 15 | 2339 | 87 | |
XM_011534146.2 | XP_011532448.1 | 19 - 20 | 3172 | 87 |