The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080423.3 | NP_001073892.3 | 21 - 22 | 2717 | 142 |
XM_011534141.2 | XP_011532443.1 | 21 - 22 | 2799 | 142 | |
XM_011534142.2 | XP_011532444.1 | 21 - 22 | 2711 | 142 | |
XM_011534143.2 | XP_011532445.1 | 20 - 21 | 2673 | 142 | |
XM_011534145.2 | XP_011532447.1 | 15 - 16 | 2445 | 142 | |
XM_011534146.2 | XP_011532448.1 | 20 - 21 | 3278 | 142 | |
GenBank mRNA | AB051506.2 | - | 21 - 22 | 2703 | 142 |
AK024507.1 | - | 8 - 9 | 1339 | 142 |