The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207359.2 | NP_997242.2 | 5 - 6 | 583 | 149 |
XM_017006297.1 | XP_016861786.1 | 5 - 6 | 981 | 149 | |
GenBank mRNA | AL832766.1 | - | 5 - 6 | 583 | 149 |