Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GRIP2
Entrez Gene ID
80852
Gene Name
glutamate receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 14489111 - 14602996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK024507.1 - 7 - 8 950 62

View Full Product Details