The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159927.1 | NP_001153399.1 | 6 - 7 | 780 | 90 |
NM_001159929.1 | NP_001153401.1 | 6 - 7 | 789 | 90 | |
NM_207102.2 | NP_996985.2 | 7 - 8 | 963 | 90 | |
XM_017006224.1 | XP_016861713.1 | 6 - 7 | 976 | 90 | |
GenBank mRNA | AY247969.1 | - | 7 - 8 | 963 | 90 |
BC101303.1 | - | 6 - 7 | 1136 | 90 | |
BC101304.1 | - | 6 - 7 | 724 | 90 | |
BC101305.1 | - | 6 - 7 | 733 | 90 | |
BC101306.1 | - | 4 - 5 | 471 | 90 |