The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020759.2 | NP_065810.2 | 5 - 6 | 437 | 72 |
XM_011521831.2 | XP_011520133.1 | 5 - 6 | 450 | 72 | |
XM_011521832.2 | XP_011520134.1 | 5 - 6 | 447 | 72 | |
XM_011521834.2 | XP_011520136.1 | 5 - 6 | 450 | 72 | |
XM_011521835.2 | XP_011520137.1 | 5 - 6 | 450 | 72 | |
XM_011521836.2 | XP_011520138.1 | 5 - 6 | 752 | 72 | |
XM_011521837.2 | XP_011520139.1 | 3 - 4 | 219 | 72 | |
XM_017022439.1 | XP_016877928.1 | 5 - 6 | 450 | 72 | |
XM_017022440.1 | XP_016877929.1 | 4 - 5 | 349 | 72 | |
XM_017022441.1 | XP_016877930.1 | 3 - 4 | 668 | 72 | |
GenBank mRNA | AK122666.1 | - | 3 - 4 | 451 | 72 |
CR627426.1 | - | 5 - 6 | 437 | 72 |