Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC22A20
Entrez Gene ID
440044
Gene Name
solute carrier family 22 member 20
Gene Aliases
Oat6
Chromosome Location
Chr.11: 65213840 - 65243108 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_033396.1 8 - 9 1406 86
XM_017017774.1 XP_016873263.1 7 - 8 1237 86
XM_017017775.1 XP_016873264.1 5 - 6 1213 86
XM_017017776.1 XP_016873265.1 5 - 6 1213 86
GenBank mRNA AK094565.1 - 1 - 2 106 86
DN831898.1 - 1 - 2 86
DN831899.1 - 4 - 5 501 86

View Full Product Details