Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A10
Entrez Gene ID
387775
Gene Name
solute carrier family 22 member 10
Gene Aliases
OAT5, hOAT5
Chromosome Location
Chr.11: 63268257 - 63311983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039752.3 NP_001034841.3 2 - 3 721 67
NR_134874.1 3 - 4 476 67
XM_011545016.2 XP_011543318.1 4 - 5 1726 67
XM_017017699.1 XP_016873188.1 3 - 4 523 67
XM_017017701.1 XP_016873190.1 4 - 5 1726 67
XM_017017702.1 XP_016873191.1 4 - 5 1726 67
GenBank mRNA AB075876.1 - 3 - 4 476 67
AK298430.1 - 3 - 4 523 67
DR003989.1 - 2 - 3 255 67

View Full Product Details