Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC22A10
Entrez Gene ID
387775
Gene Name
solute carrier family 22 member 10
Gene Aliases
OAT5, hOAT5
Chromosome Location
Chr.11: 63268257 - 63311983 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039752.3 NP_001034841.3 8 - 9 1601 114
NR_134874.1 9 - 10 1217 114
XM_011545016.2 XP_011543318.1 10 - 11 2606 114
XM_017017699.1 XP_016873188.1 9 - 10 1403 114
XM_017017700.1 XP_016873189.1 8 - 9 1839 114
XM_017017701.1 XP_016873190.1 10 - 11 2467 114
XM_017017703.1 XP_016873192.1 7 - 8 947 114
XM_017017704.1 XP_016873193.1 8 - 9 1699 114
GenBank mRNA AB075876.1 - 9 - 10 1217 114
AK298430.1 - 9 - 10 1264 114
BC171744.1 - 6 - 7 797 114

View Full Product Details