Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAMB4
Entrez Gene ID
22798
Gene Name
laminin subunit beta 4
Gene Aliases
-
Chromosome Location
Chr.7: 108011662 - 108130356 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318046.1 NP_001304975.1 13 - 14 1633 71
NM_001318047.1 NP_001304976.1 13 - 14 1636 71
NM_001318048.1 NP_001304977.1 13 - 14 1633 71
NM_007356.2 NP_031382.2 13 - 14 1636 71
XM_011515975.1 XP_011514277.1 13 - 14 1636 71
XM_011515978.1 XP_011514280.1 13 - 14 1636 71
XM_011515979.2 XP_011514281.1 13 - 14 1636 71
XM_011515980.2 XP_011514282.1 13 - 14 1636 71
XM_017011879.1 XP_016867368.1 13 - 14 1636 71
XM_017011880.1 XP_016867369.1 13 - 14 1636 71
GenBank mRNA AF028816.1 - 13 - 14 1632 71
AK300385.1 - 13 - 14 1637 71
BC140804.1 - 13 - 14 1623 71
BC142619.1 - 13 - 14 1618 71
BC144591.1 - 13 - 14 1623 71

View Full Product Details