Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- SFTA1P
- Entrez Gene ID
-
207107
- Gene Name
- surfactant associated 1, pseudogene
- Gene Aliases
-
SFTPF
- Chromosome Location
-
Chr.10: 10784439 - 10794914 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_027082.1 |
|
1 - 2 |
217 |
104 |
GenBank mRNA |
AY102069.1 |
- |
1 - 2 |
217 |
104 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
T96554.1 |
- |
|
136 |
|
Chr. - - [-] on Build |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101928322 |
LOC101928322 |
|
-330.0kb
|
Sense |
Chr. - 10419538 - 10462349 on Build GRCh38 |
Downstream |
long intergenic non-protein coding RNA 710 |
LINC00710 |
|
140.0kb
|
Sense |
Chr. - 10934940 - 10952163 on Build GRCh38 |
View Full Product Details