Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A25
Entrez Gene ID
387601
Gene Name
solute carrier family 22 member 25
Gene Aliases
HIMTP, UST6
Chromosome Location
Chr.11: 63162264 - 63243523 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017017684.1 XP_016873173.1 5 - 6 1581 66
XM_017017685.1 XP_016873174.1 6 - 7 1709 66
XM_017017686.1 XP_016873175.1 5 - 6 2017 66
XM_017017689.1 XP_016873178.1 1 - 2 728 66
XM_017017695.1 XP_016873184.1 5 - 6 2122 66
GenBank mRNA AK091990.1 - 1 - 2 50 66
AY517500.1 - 1 - 2 50 66

View Full Product Details