The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135914.1 | NP_001129386.1 | 16 - 17 | 1748 | 112 |
NM_199349.2 | NP_955381.2 | 18 - 19 | 1922 | 112 | |
XM_017012184.1 | XP_016867673.1 | 18 - 19 | 1922 | 112 | |
XM_017012185.1 | XP_016867674.1 | 18 - 19 | 1922 | 112 | |
XM_017012186.1 | XP_016867675.1 | 18 - 19 | 1922 | 112 | |
XM_017012187.1 | XP_016867676.1 | 18 - 19 | 1922 | 112 | |
XM_017012188.1 | XP_016867677.1 | 18 - 19 | 1922 | 112 | |
XM_017012189.1 | XP_016867678.1 | 18 - 19 | 1922 | 112 | |
XM_017012190.1 | XP_016867679.1 | 18 - 19 | 1922 | 112 | |
XM_017012191.1 | XP_016867680.1 | 11 - 12 | 1141 | 112 | |
XM_017012192.1 | XP_016867681.1 | 11 - 12 | 1152 | 112 | |
XM_017012193.1 | XP_016867682.1 | 18 - 19 | 1922 | 112 | |
XM_017012194.1 | XP_016867683.1 | 18 - 19 | 1922 | 112 | |
GenBank mRNA | AK095464.1 | - | 11 - 12 | 1396 | 112 |
AK122706.1 | - | 18 - 19 | 1922 | 112 |