Product Details

Assay Reference Genome
Location

Chr.10:93757934 on build GRCh38
Cytoband
10q23.33
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
9211
Gene Symbol
LGI1
Gene Name
leucine rich glioma inactivated 1
Gene Aliases
ADLTE, ADPAEF, ADPEAF, EPITEMPIN, EPT, ETL1, IB1099
Location
Chr.10:93757770-93798174 on build GRCh38
Assay Gene Location
Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
LGI1 NM_001308275.1 1 126 NP_001295204.1
NM_001308276.1 1 126 NP_001295205.1
NM_005097.3 1 126 NP_005088.1
NR_131777.1 1 126
XM_017016911.1 1 59 XP_016872400.1
XM_017016912.1 1 165 XP_016872401.1
AF055636.1 AAC99316.1
AF473548.1 AAM22074.1
AK303956.1
AK309416.1
AY358885.1 AAQ89244.1
BC022500.1 1 50 AAH22500.1
DC326684.1 1 126

View Full Product Details