The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001321708.1 | NP_001308637.1 | 29 - 30 | 2798 | 120 |
NM_001321709.1 | NP_001308638.1 | 30 - 31 | 2576 | 120 | |
NM_004717.3 | NP_004708.1 | 30 - 31 | 2822 | 120 | |
XM_011516682.2 | XP_011514984.2 | 30 - 31 | 2858 | 120 | |
XM_011516684.2 | XP_011514986.2 | 23 - 24 | 10042 | 120 | |
XM_017012780.1 | XP_016868269.1 | 30 - 31 | 2859 | 120 | |
XM_017012781.1 | XP_016868270.1 | 30 - 31 | 2843 | 120 | |
XM_017012782.1 | XP_016868271.1 | 29 - 30 | 2744 | 120 | |
XM_017012784.1 | XP_016868273.1 | 28 - 29 | 2669 | 120 | |
XM_017012785.1 | XP_016868274.1 | 28 - 29 | 3046 | 120 | |
XM_017012786.1 | XP_016868275.1 | 30 - 31 | 2546 | 120 | |
XM_017012787.1 | XP_016868276.1 | 29 - 30 | 2411 | 120 | |
XM_017012788.1 | XP_016868277.1 | 30 - 31 | 2529 | 120 | |
XM_017012789.1 | XP_016868278.1 | 16 - 17 | 1255 | 120 |