Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA3
Entrez Gene ID
2802
Gene Name
golgin A3
Gene Aliases
GCP170, MEA-2
Chromosome Location
Chr.12: 132768909 - 132829131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172557.1 NP_001166028.1 12 - 13 2770 118
NM_005895.3 NP_005886.2 12 - 13 3108 118
XM_005266164.4 XP_005266221.1 12 - 13 2715 118
XM_005266167.4 XP_005266224.1 9 - 10 2140 118
XM_006719736.3 XP_006719799.1 12 - 13 2835 118
XM_006719737.3 XP_006719800.1 12 - 13 2845 118
XM_011534793.2 XP_011533095.1 11 - 12 2562 118
XM_011534794.2 XP_011533096.1 4 - 5 713 118
GenBank mRNA AB027133.1 - 12 - 13 2830 118
AF485338.1 - 11 - 12 2552 118
BC142658.1 - 12 - 13 2770 118
BC146675.1 - 12 - 13 2770 118
D63997.1 - 12 - 13 2818 118
L06148.1 - 2 - 3 242 118

View Full Product Details