The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172557.1 | NP_001166028.1 | 13 - 14 | 3033 | 61 |
NM_005895.3 | NP_005886.2 | 13 - 14 | 3371 | 61 | |
XM_005266164.4 | XP_005266221.1 | 13 - 14 | 2978 | 61 | |
XM_005266167.4 | XP_005266224.1 | 10 - 11 | 2403 | 61 | |
XM_006719736.3 | XP_006719799.1 | 13 - 14 | 3098 | 61 | |
XM_006719737.3 | XP_006719800.1 | 13 - 14 | 3108 | 61 | |
XM_011534793.2 | XP_011533095.1 | 12 - 13 | 2825 | 61 | |
XM_011534794.2 | XP_011533096.1 | 5 - 6 | 976 | 61 | |
GenBank mRNA | AB027133.1 | - | 13 - 14 | 3093 | 61 |
BC142658.1 | - | 13 - 14 | 3033 | 61 | |
BC146675.1 | - | 13 - 14 | 3033 | 61 | |
D63997.1 | - | 13 - 14 | 3081 | 61 | |
L06148.1 | - | 3 - 4 | 505 | 61 |