Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GOLGA3
Entrez Gene ID
2802
Gene Name
golgin A3
Gene Aliases
GCP170, MEA-2
Chromosome Location
Chr.12: 132768909 - 132829131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005895.3 NP_005886.2 20 - 21 4416 59
XM_005266164.4 XP_005266221.1 20 - 21 4023 59
XM_005266167.4 XP_005266224.1 17 - 18 3448 59
XM_006719736.3 XP_006719799.1 20 - 21 4143 59
XM_006719737.3 XP_006719800.1 20 - 21 4153 59
XM_011534793.2 XP_011533095.1 19 - 20 3870 59
XM_011534794.2 XP_011533096.1 12 - 13 2021 59
GenBank mRNA AB027133.1 - 20 - 21 4138 59
D63997.1 - 20 - 21 4126 59
L06148.1 - 10 - 11 1550 59

View Full Product Details