The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172557.1 | NP_001166028.1 | 5 - 6 | 1402 | 72 |
NM_005895.3 | NP_005886.2 | 5 - 6 | 1740 | 72 | |
XM_005266164.4 | XP_005266221.1 | 5 - 6 | 1347 | 72 | |
XM_005266167.4 | XP_005266224.1 | 2 - 3 | 772 | 72 | |
XM_006719736.3 | XP_006719799.1 | 5 - 6 | 1467 | 72 | |
XM_006719737.3 | XP_006719800.1 | 5 - 6 | 1477 | 72 | |
XM_011534793.2 | XP_011533095.1 | 4 - 5 | 1194 | 72 | |
GenBank mRNA | AB027133.1 | - | 5 - 6 | 1462 | 72 |
AF485338.1 | - | 4 - 5 | 1184 | 72 | |
BC060826.1 | - | 5 - 6 | 1426 | 72 | |
BC142658.1 | - | 5 - 6 | 1402 | 72 | |
BC146675.1 | - | 5 - 6 | 1402 | 72 | |
D63997.1 | - | 5 - 6 | 1450 | 72 |