Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLGA3
Entrez Gene ID
2802
Gene Name
golgin A3
Gene Aliases
GCP170, MEA-2
Chromosome Location
Chr.12: 132768909 - 132829131 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172557.1 NP_001166028.1 8 - 9 2022 89
NM_005895.3 NP_005886.2 8 - 9 2360 89
XM_005266164.4 XP_005266221.1 8 - 9 1967 89
XM_005266167.4 XP_005266224.1 5 - 6 1392 89
XM_006719736.3 XP_006719799.1 8 - 9 2087 89
XM_006719737.3 XP_006719800.1 8 - 9 2097 89
XM_011534793.2 XP_011533095.1 7 - 8 1814 89
GenBank mRNA AB027133.1 - 8 - 9 2082 89
AF485338.1 - 7 - 8 1804 89
BC060826.1 - 8 - 9 2046 89
BC142658.1 - 8 - 9 2022 89
BC146675.1 - 8 - 9 2022 89
D63997.1 - 8 - 9 2070 89

View Full Product Details