The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006665.1 | NP_001006666.1 | 20 - 21 | 2163 | 92 |
NM_002953.3 | NP_002944.2 | 21 - 22 | 2250 | 92 | |
XM_005245966.4 | XP_005246023.1 | 20 - 21 | 2194 | 92 | |
GenBank mRNA | AK092955.1 | - | 21 - 22 | 2252 | 92 |
AK292722.1 | - | 20 - 21 | 2165 | 92 | |
AK299007.1 | - | 20 - 21 | 2019 | 92 | |
AK315730.1 | - | 21 - 22 | 2166 | 92 | |
BC014966.1 | - | 21 - 22 | 2243 | 92 | |
L07597.1 | - | 21 - 22 | 2133 | 92 |