Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ANAPC4
Entrez Gene ID
29945
Gene Name
anaphase promoting complex subunit 4
Gene Aliases
APC4
Chromosome Location
Chr.4: 25377226 - 25418498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286756.1 NP_001273685.1 6 - 7 596 102
NM_013367.2 NP_037499.2 6 - 7 596 102
XM_011513838.1 XP_011512140.1 6 - 7 473 102
XM_011513839.1 XP_011512141.1 6 - 7 517 102
XM_017008138.1 XP_016863627.1 6 - 7 517 102
GenBank mRNA AF191338.1 - 5 - 6 467 102
AK023597.1 - 3 - 4 207 102
AK292336.1 - 6 - 7 516 102
BC059383.1 - 6 - 7 569 102

View Full Product Details