The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031421.3 | NP_113609.1 | 5 - 6 | 730 | 101 |
XM_011525323.2 | XP_011523625.1 | 5 - 6 | 670 | 101 | |
XM_011525324.2 | XP_011523626.1 | 5 - 6 | 697 | 101 | |
GenBank mRNA | AK055498.1 | - | 5 - 6 | 675 | 101 |
AL136760.1 | - | 5 - 6 | 697 | 101 | |
BC067297.1 | - | 5 - 6 | 689 | 101 | |
CF593846.1 | - | 5 - 6 | 668 | 101 |