The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006875.3 | NP_006866.2 | 1 - 2 | 368 | 144 |
GenBank mRNA | AK290931.1 | - | 1 - 2 | 256 | 144 |
AK312745.1 | - | 1 - 2 | 256 | 144 | |
BC018111.1 | - | 1 - 2 | 239 | 144 | |
DC378253.1 | - | 1 - 2 | 368 | 144 | |
U77735.1 | - | 1 - 2 | 251 | 144 |