The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267843.1 | NP_001254772.1 | 14 - 15 | 1906 | 95 |
NM_004898.3 | NP_004889.1 | 13 - 14 | 1397 | 95 | |
XM_005265787.2 | XP_005265844.1 | 13 - 14 | 1443 | 95 | |
XM_011534409.2 | XP_011532711.1 | 13 - 14 | 1852 | 95 | |
XM_011534410.2 | XP_011532712.1 | 12 - 13 | 1234 | 95 | |
XM_011534411.2 | XP_011532713.1 | 12 - 13 | 1193 | 95 | |
XM_017008854.1 | XP_016864343.1 | 14 - 15 | 1783 | 95 | |
XM_017008855.1 | XP_016864344.1 | 10 - 11 | 992 | 95 | |
GenBank mRNA | AB002332.1 | - | 12 - 13 | 1231 | 95 |
AB005535.1 | - | 13 - 14 | 1601 | 95 | |
AF011568.1 | - | 13 - 14 | 1337 | 95 | |
AK223549.1 | - | 13 - 14 | 1337 | 95 | |
AK291708.1 | - | 13 - 14 | 1336 | 95 | |
BC041878.1 | - | 14 - 15 | 1906 | 95 | |
BC126157.1 | - | 13 - 14 | 1313 | 95 | |
BC126159.1 | - | 13 - 14 | 1313 | 95 |