The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002249.5 | NP_002240.3 | 3 - 4 | 1763 | 68 |
NM_170782.2 | NP_740752.1 | 3 - 4 | 684 | 68 | |
GenBank mRNA | AF438203.1 | - | 3 - 4 | 662 | 68 |
AJ251016.1 | - | 3 - 4 | 1797 | 68 | |
AY049734.1 | - | 3 - 4 | 1534 | 68 | |
AY138900.1 | - | 3 - 4 | 887 | 68 | |
AY258142.1 | - | 3 - 4 | 1467 | 68 | |
BC042147.1 | - | 3 - 4 | 665 | 68 | |
Y08263.1 | - | 3 - 4 | 1738 | 68 |