The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174081.1 | NP_001167552.1 | 6 - 7 | 2145 | 59 |
NM_001174082.1 | NP_001167553.1 | 5 - 6 | 2110 | 59 | |
NM_001174083.1 | NP_001167554.1 | 7 - 8 | 2107 | 59 | |
NM_017852.3 | NP_060322.1 | 6 - 7 | 2176 | 59 | |
GenBank mRNA | AB209300.1 | - | 5 - 6 | 2028 | 59 |
AF298547.1 | - | 5 - 6 | 2027 | 59 | |
AF310106.1 | - | 6 - 7 | 2121 | 59 | |
AF464764.1 | - | 6 - 7 | 2076 | 59 | |
AK000517.1 | - | 6 - 7 | 2121 | 59 | |
AK000784.1 | - | 1 - 2 | 296 | 59 | |
AK025952.1 | - | 6 - 7 | 2085 | 59 | |
AK223253.1 | - | 6 - 7 | 2085 | 59 | |
AK223269.1 | - | 6 - 7 | 2085 | 59 | |
AK292681.1 | - | 6 - 7 | 2104 | 59 | |
AK302989.1 | - | 7 - 8 | 2076 | 59 | |
BC001039.2 | - | 5 - 6 | 2056 | 59 | |
BC003592.1 | - | 6 - 7 | 2100 | 59 | |
BC039269.1 | - | 6 - 7 | 2145 | 59 |