Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLRP7
Entrez Gene ID
199713
Gene Name
NLR family pyrin domain containing 7
Gene Aliases
CLR19.4, HYDM, NALP7, NOD12, PAN7, PYPAF3
Chromosome Location
Chr.19: 54923509 - 54965184 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127255.1 NP_001120727.1 1 - 2 38 65
NM_139176.3 NP_631915.2 1 - 2 38 65
NM_206828.3 NP_996611.2 1 - 2 38 65
XM_006723075.3 XP_006723138.1 2 - 3 273 65
XM_011526596.2 XP_011524898.1 3 - 4 480 65
XM_011526599.2 XP_011524901.1 3 - 4 491 65
XM_011526601.2 XP_011524903.1 3 - 4 480 65
GenBank mRNA AK298630.1 - 1 - 2 55 65
AK302655.1 - 3 - 4 449 65
AK313964.1 - 3 - 4 433 65
AY154462.1 - 1 - 2 32 65
BC109124.2 - 1 - 2 30 65
DC425389.1 - 1 - 2 55 65

View Full Product Details