The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127255.1 | NP_001120727.1 | 4 - 5 | 2006 | 83 |
NM_206828.3 | NP_996611.2 | 4 - 5 | 2006 | 83 | |
XM_006723075.3 | XP_006723138.1 | 5 - 6 | 2241 | 83 | |
XM_006723076.3 | XP_006723139.1 | 4 - 5 | 2204 | 83 | |
XM_011526596.2 | XP_011524898.1 | 6 - 7 | 2448 | 83 | |
XM_011526599.2 | XP_011524901.1 | 6 - 7 | 2459 | 83 | |
XM_011526601.2 | XP_011524903.1 | 6 - 7 | 2448 | 83 | |
GenBank mRNA | AF464765.1 | - | 3 - 4 | 1976 | 83 |
AK298630.1 | - | 3 - 4 | 1324 | 83 | |
AK302655.1 | - | 6 - 7 | 2417 | 83 | |
AK313964.1 | - | 6 - 7 | 2401 | 83 | |
AY154462.1 | - | 4 - 5 | 2000 | 83 | |
BC109124.2 | - | 4 - 5 | 1998 | 83 | |
BC109125.1 | - | 4 - 5 | 1998 | 83 |