The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318509.1 | NP_001305438.1 | 14 - 15 | 2319 | 127 |
NM_001318510.1 | NP_001305439.1 | 14 - 15 | 2028 | 127 | |
NM_004458.2 | NP_004449.1 | 14 - 15 | 2014 | 127 | |
NM_022977.2 | NP_075266.1 | 15 - 16 | 2326 | 127 | |
XM_005262109.2 | XP_005262166.1 | 13 - 14 | 2188 | 127 | |
XM_006724635.2 | XP_006724698.1 | 13 - 14 | 1806 | 127 | |
XM_011530888.2 | XP_011529190.1 | 14 - 15 | 2068 | 127 | |
XM_011530889.2 | XP_011529191.1 | 13 - 14 | 2074 | 127 | |
GenBank mRNA | AB061713.1 | - | 14 - 15 | 1908 | 127 |
AB061714.1 | - | 14 - 15 | 1868 | 127 | |
AF030555.1 | - | 14 - 15 | 2015 | 127 | |
AK292070.1 | - | 14 - 15 | 1910 | 127 | |
AK294915.1 | - | 14 - 15 | 1998 | 127 | |
BC034959.2 | - | 14 - 15 | 1877 | 127 | |
Y12777.1 | - | 14 - 15 | 1878 | 127 |