The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177880.1 | NP_001171351.1 | 2 - 3 | 260 | 81 |
NM_177963.3 | NP_808878.1 | 2 - 3 | 169 | 81 | |
XM_006718737.3 | XP_006718800.1 | 4 - 5 | 1020 | 81 | |
XM_011545346.2 | XP_011543648.1 | 5 - 6 | 1192 | 81 | |
XM_011545347.1 | XP_011543649.1 | 2 - 3 | 110 | 81 | |
GenBank mRNA | AK024280.1 | - | 5 - 6 | 1192 | 81 |
AK289672.1 | - | 2 - 3 | 122 | 81 | |
AK289727.1 | - | 2 - 3 | 122 | 81 | |
AK315810.1 | - | 2 - 3 | 261 | 81 | |
BC037406.1 | - | 2 - 3 | 94 | 81 | |
DA239473.1 | - | 2 - 3 | 169 | 81 | |
HY172636.1 | - | 2 - 3 | 261 | 81 |